Skip to Content
Merck
All Photos(1)

Key Documents

818074

TOYOPEARL® AF-Amino-650M Bulk Media

phase amine-activated, bottle of 1 L, 65 μm particle size

Synonym(s):

TOYOPEARL® AF-Amino-650M Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
particle size:
65 μm
matrix active group:
amine, primary
amine-activated phase
pore size:
1000 Å
matrix:
hydroxylated methacrylic polymer particle platform
Pricing and availability is not currently available.

description

Adsorption Capacity (> 80 g/L IgG (static); >70 g/L IgG (dynamic at 5 min. residence time))
Clean in Place (CIP) (0.1 M NaOH)

product line

TOYOPEARL®

form

slurry

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 18074

extent of labeling

100 μmol/mL±30 ligand density

parameter

pressure (max. 3 bar)

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer particle platform

matrix active group

amine, primary
amine-activated phase

particle size

65 μm

pore size

1000 Å

pH

2-13

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Columns are tolerant to aqueous buffers and salt solutions, 1 mol/L alkaline solutions, 1 mol/L non-oxidizing mineral acids, 8 mol/L urea, 6 mol/L guanidine hydrochloride, non-halogenated organic solvents and detergents. They are not compatible with strong oxidants and halogenated solvents.
Toyopearl AF-Amino-650 resin is a reactive resin used for the coupling of specific ligands for affinity chromatography. Ligands are immobilized by either peptide bond formation or reductive amination through their respective carboxylate or aldehyde groups.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service