Skip to Content
Merck
All Photos(1)

Documents

814695

TOYOPEARL® CM-650S Bulk Media

phase carboxymethyl, bottle of 1 L, 35 μm particle size

Synonym(s):

TOYOPEARL® CM-650 Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

product line

TOYOPEARL®

form

slurry

feature

counter ion Na+

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 14695

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

carboxymethyl phase

particle size

35 μm

operating pH

2-13

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl CM-650 resin is a weak cation exchanger for biomolecule purification. It can be used for high throughput capture, intermediate purification, and polishing process steps.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service