Skip to Content
Merck
All Photos(1)

Key Documents

808802

TSKgel® DEAE-5PW (10 µm) HPLC Columns

L x I.D. 7.5 cm × 8 mm, DEAE (diethylaminoethyl) phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
Pricing and availability is not currently available.

Product Name

TSKgel® DEAE-5PW HPLC Column, phase DEAE (diethylaminoethyl), L × I.D. 7.5 cm × 8 mm, 10 μm particle size

material

glass

product line

TSKgel®

feature

counter ion Cl-

manufacturer/tradename

Tosoh 8802

parameter

≤1 M salt concentration
≤20% organic solvent
1.2 mL/min flow rate
10 bar max. pressure
10-45 °C temperature

technique(s)

HPLC: suitable

L × I.D.

7.5 cm × 8 mm

matrix

hydroxylated methacrylic polymer

matrix active group

DEAE (diethylaminoethyl) phase

particle size

10 μm

operating pH

2-12

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel® DEAE-5PW columns and guard columns are packed with porous hydrophilic polymer particles which are modified with a weak anion exchanger. They are used for the separation and analysis of proteins, nucleotides, nucleosides and other biologically active molecules. Columns are available in internal diameters varying from 2mm to 55mm ID. Column housings are either glass or stainless steel. They have the same backbone chemistry and selectivity as the bulk process scale resins Toyopearl® DEAE-650 and TSKgel DEAE-5PW (20) and (30).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation
Toyopearl is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service