Skip to Content
Merck
All Photos(1)

Documents

808544

TSKgel® SWXL (5 µm) HPLC Columns

bottle of 1g, diol phase

Synonym(s):

TSKgel® SWXL Columns, TSKgel® SWxl Top-Off, 5μm, for TSKgel® SWxl and QC-PAK columns, 1g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

product name

TSKgel® SWXL Top-Off Gel, phase diol, 5 μm particle size, bottle of 1 g

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

form

slurry

packaging

bottle of 1 g

manufacturer/tradename

Tosoh 08544

parameter

≤0.5 M salt concentration
0-100% aqueous soluble organic solvents
10-30 °C temperature

technique(s)

HPLC: suitable

matrix

silica particle platform

matrix active group

diol phase

particle size

5 μm

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Top-Off for TSKgel SWxl and QC-PAK columns, 5 μm, 1 gram

Application

TSKgel® SWXL top-off gel may be used as a silica-based support column in the separation of proteins using high performance liquid chromatography (HPLC).

Physical form

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

102.2 °F

Flash Point(C)

39 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New high-performance gel filtration columns for protein separation.
Kato Y, et al.
Journal of Chromatography A, 404, 333-339 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service