Skip to Content
Merck
All Photos(1)

Documents

807124

TSKgel® ODS-120A (5 µm) HPLC Columns

L x I.D. 25 cm × 4.6 mm, C18 (octadecyl) phase

Synonym(s):

TSKgel® Reversed Phase HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

TSKgel® ODS-120A HPLC Column, phase C18 (octadecyl), L × I.D. 25 cm × 4.6 mm, 5 μm particle size

material

stainless steel

Agency

suitable for USP L1

product line

TSKgel®

manufacturer/tradename

Tosoh ODS-120A, 07124

extent of labeling

20% Carbon, multilayer, not encapped

parameter

0-100% organic solvent
0-45 °C temperature
1.2 mL/min flow rate
200 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

200 m2/g

matrix

silica particle platform

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

150 Å pore diameter

operating pH

2.0-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel ODS-120A columns use a 150Å pore size silica base support. The silica′s exclusion limit of 10,000 Da makes them a good choice for the reversed phase chromatography of peptides, small proteins, and environmental samples such as poly-aromatic hydrocarbons. The "A" signifies that the material is not endcapped. For charged samples, the endcapped TSKgel ODS-120T column is a more suitable alternative. Bonded phase pore size is indicated by the number in the product description, in this case TSKgel ODS-120A has 120Å nominal pore size.

Physical form

Shipped in 70% methanol - 30% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service