Skip to Content
Merck
All Photos(1)

Documents

713-0141

HPLC Analysis of Barbiturates on Discovery® C18

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Discovery C18, 15 cm × 4.6 mm I.D., 5 μm particles (504955)
mobile phase: [A] acetonitrile; [B] 25 mM potassium phosphate buffer, pH 6.9 (25:75, A:B)
flow rate: 1.5 mL/min
column temp.: ambient
detector: UV, 254 nm
injection: 25 μL

suitability

application for HPLC

application(s)

forensics and toxicology
pharmaceutical (small molecule)

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Methyprylon
    Hexobarbital
    Butalbital
    Butabarbital

analytical column


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service