Skip to Content
Merck
All Photos(1)

Key Documents

67243-U

Supelco

BIOshell A160 Peptide C18 (2 µm) HPLC Columns

L × I.D. 15 cm × 2.1 mm, HPLC Column

Synonym(s):

Superficially Porous Wide Pore C18 UHPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52
Pricing and availability is not currently available.

Product Name

BIOshell A160 Peptide C18 Column, 2 µm HPLC Column, 2.0 μm particle size, L × I.D. 15 cm × 2.1 mm

material

stainless steel hardware

Quality Level

Agency

suitable for USP L1

product line

BIOshell

feature

endcapped: no

analyte chemical class(es)

peptides

manufacturer/tradename

BIOshell

packaging

1 ea of

parameter

14,500 psi max. pressure (1000 bar)
60 °C max. temp.
600 bar pressure (8702 psi)

technique(s)

HPLC: suitable
LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable

L × I.D.

15 cm × 2.1 mm

surface area

65 m2/g

matrix

spherical silica particle platform
superficially porous particle

matrix active group

C18 (octadecyl) phase

particle size

2.0 μm

pore size

160 Å pore size

operating pH range

1-8

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Shell thickness: 0.4 μm
Solid Core: 1.2 μm
Extent of labeling: 4.0%

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Method development for protein fingerprinting of AAV serotype 5 using both intact mass analysis and peptide mapping, to determine critical quality attributes for gene therapy, utilizing three different columns.

Related Content

HPLC separates and identifies large biomolecules like proteins and peptides by pumping the sample through a sorbent-packed column.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service