Documents
59262-U
SSI High Pressure Preinjector Filter
stainless steel
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
material
stainless steel
packaging
pk of 1 ea
General description
Place between the pump and injector to provide final filtration for the mobile phase. Includes 316 stainless steel filter element (0.5 μm pores) that is easy to replace. Maximum operating pressure: 15,000 psi (105 MPa). For 1/16 in. O.D. tubing, 10-32 threads.
Legal Information
SSI is a trademark of Scientific Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Articles
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.