Skip to Content
Merck
All Photos(1)

Key Documents

59200-U

Supelco

Postcolumn Reactor Components, Glass Beads

size 75 μm, acid-washed

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
SB.51
Pricing and availability is not currently available.

packaging

pkg of 25 g

size

75 μm

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Po-Hsiang Tsui et al.
Scientific reports, 7, 41004-41004 (2017-01-21)
Constructing ultrasound statistical parametric images by using a sliding window is a widely adopted strategy for characterizing tissues. Deficiency in spatial resolution, the appearance of boundary artifacts, and the prerequisite data distribution limit the practicability of statistical parametric imaging. In
Zhuhuang Zhou et al.
Ultrasonic imaging, 40(3), 171-189 (2018-03-07)
In this study, the window-modulated compounding (WMC) technique was integrated into three-dimensional (3D) ultrasound Nakagami imaging for improving the spatial visualization of backscatter statistics. A 3D WMC Nakagami image was produced by summing and averaging a number of 3D Nakagami
Xue Yu et al.
Physics in medicine and biology, 60(19), 7513-7531 (2015-09-16)
Ultrasound tissue characterization is crucial for the detection of tissue abnormalities. Since the statistics of the backscattered ultrasound signals strongly depend on density and spatial arrangement of local scatterers, appropriate modeling of the backscattered signals may be capable of providing
Jui Fang et al.
Ultrasonics, 87, 91-102 (2018-02-25)
Hepatic steatosis is an abnormal state where excess lipid mass is accumulated in hepatocyte vesicles. Backscattered ultrasound signals received from the liver contain useful information regarding the degree of steatosis in the liver. The homodyned-K (HK) distribution has been demonstrated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service