Skip to Content
Merck
All Photos(1)

Documents

57661

Tee

bore 0.020 in., 10-32 thread

Synonym(s):

Upchurch PEEK Fingertight Union

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815

bore

0.020 in.

fitting

10-32 thread

Looking for similar products? Visit Product Comparison Guide

General description

The PEEK tees are to be operated at 4000 ψ (281 kg/cm2). They include Fingertight III nuts and ferrules.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

András Domokos et al.
International journal of pharmaceutics, 581, 119297-119297 (2020-04-04)
An end-to-end continuous pharmaceutical manufacturing process was developed for the production of conventional direct compressed tablets on a proof-of-concept level for the first time. The output reaction mixture of the flow synthesis of acetylsalicylic acid was crystallized continuously in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service