Key Documents
57630
Sample Loop for Model 7725 Injector
volume 1 mL
About This Item
UNSPSC Code:
41115854
Pricing and availability is not currently available.
material
stainless steel sample loop and fittings
manufacturer/tradename
Rheodyne 7755-027
volume
1 mL
compatibility
for use with Rheodyne 7725 and 7725i injector
General description
Sample loop for model 7725 injector is designed for advanced fluid-handling solutions for a wide range of analytical instrumentation and in vitro diagnostic systems. It can be used in high performance liquid chromatographic systems and is quite accurate and precise in injecting 1μL-5 mL concentration of samples.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Articles
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.