Skip to Content
Merck
All Photos(1)

Documents

57584-U

Supelco

Diffusive Sampling Fiber Holder for SPME FFA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
NB.21

technique(s)

solid phase microextraction (SPME): suitable

application(s)

food and beverages

General description

A holder for SPME FFA capable of determining the time-weighted average (TWA) concentration of volatile organic compounds (VOCs) in air. Unlike conventional sampling with SPME in which the fiber is extended outside the needle, during TWAS passive sampling the fiber is retracted a known distance inside the needle. The sample collect VOCs by the mechanism of molecular diffusion and sorption onto the fiber.

Other Notes

Although we no longer distribute SPME-FFA products andaccessories, these products are available and distributed exclusively byChromline. Please visit the Chromline websiteor contact info@chromlinesrl.com for more information.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola Mucci et al.
La Medicina del lavoro, 110(6), 446-458 (2019-12-18)
In recent years, under-vacuum sealing (UVS) and containers with formalin encapsulated in the lid have been proposed for the reduction of occupational exposure to airborne formaldehyde (FA) in healthcare environments. We are presenting a study focused on the assessment of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service