Skip to Content
Merck
All Photos(1)

Documents

57031-U

Supelco

Visiprep Manifold Cover (flow control valves and gasket included)

for use with standard, 12-port model

Synonym(s):

V.M.COVER W/12 CONTROL VALVES AND GASKET

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.21

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

compatibility

for use with standard, 12-port model

Looking for similar products? Visit Product Comparison Guide

Footnote

Included with Visiprep 12-port Vacuum Manifold (57030-U)

Legal Information

Visiprep is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service