56302AST
apHera™ C4 (5 μm) HPLC Columns
L × I.D. 15 cm × 4.6 mm, HPLC Column
Synonym(s):
Polymeric C4 Reversed-Phase Column
About This Item
Product Name
apHera™ C4 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
material
stainless steel column
product line
apHera™
packaging
pkg of 1 ea
manufacturer/tradename
apHera™
parameter
147 bar pressure (2133 psi)
4-60 °C temperature
technique(s)
HPLC: suitable
LC/MS: suitable
L × I.D.
15 cm × 4.6 mm
matrix
polymeric
matrix active group
C4 (butyl) phase
particle size
5 μm
pore size
300 Å
operating pH
1-14
application(s)
food and beverages
separation technique
reversed phase
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features of apHera™ Reversed Phase Columns
- Stable vinyl copolymer base
- Stable pH range 2-12
- 300 angstrom pore size
- Stable in all organic solvents
- Expanded applications: peptides, proteins
- Efficient separation of any basic substance
- Amenability to washing and alkaline solutions
Legal Information
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service