54056-U
Supelclean™ ENVI-WAX SPE Tube
bed wt. 200 mg, suitable for PFAS testing, volume 6 mL, pk of 30 ea
Synonym(s):
Supelclean™ ENVI-WAX SPE
About This Item
Recommended Products
Agency
suitable for DIN 38407-42
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA OTM-45
suitable for GB 31604.35-2016
suitable for ISO 21675 2019
suitable for ISO 25101
suitable for ISO/CEN 15968-2010
Quality Level
reg. compliance
suitable for FDA C-010.02
composition
bed wt., 200 mg
packaging
pk of 30 ea
volume
6 mL
matrix
(mixed-mode polymeric)
matrix active group
WAX phase
diameter
40-60 μm
pore size
70-90 Å pore size
application(s)
PFAS testing
environmental
separation technique
ion exchange
Looking for similar products? Visit Product Comparison Guide
General description
Applications:
- Environmental
- Extraction of acidic compounds
- PFAS
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
LC-MS method separates 32 PFAS compounds in drinking water following EPA methods 533, 537, and 537.1. EPA 8327.
Learn how method EPA 533 is used for sample collection and preparation, featuring Supelclean™ ENVI-WAX™ SPE cartridges for LC-MS analysis on an Ascentis® Express PFAS HPLC Column.
Simultaneous determination of 27 PFAS chemical compounds after enriching the water samples with SPE using a Supelclean® ENVI-WAX weak anion exchange tube.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service