Skip to Content
Merck
All Photos(1)

Documents

506419

Supelco

ASTM® D2887 Reference Gas Oil

analytical standard

Synonym(s):

RGO

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107

grade

analytical standard

Agency

ASTM® D2887,D7169

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

suitability

suitable for

application(s)

petroleum

format

neat

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

This sample is a petroleum fraction having a boiling range from 250 °F to 850 °F, evaluated in round-robin studies by the ASTM. Use this sample and the supplied ASTM boiling range consensus values to evaluate system performance.

Application

ASTM® D2887 reference gas oil has been used as a standard in the analysis of fossil-fuel liquid in order to quantitatively measure aliphatic, aromatic and polar compounds classes using normal phase high performance liquid chromatography method (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Muta. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrocarbon class analysis of coal-derived liquids using high performance liquid chromatography
Padlo.MD, et al.
Fuel Processing Technology, 49, 247-258 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service