Skip to Content
Merck
All Photos(1)

Key Documents

506052

Supelco

Iodomethane solution

certified reference material, 2000 μg/mL in methanol: water (4:1)

Synonym(s):

Methyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3I
CAS Number:
Molecular Weight:
141.94
Beilstein:
969135
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol: water (4:1)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

-10 to -25°C

SMILES string

CI

InChI

1S/CH3I/c1-2/h1H3

InChI key

INQOMBQAUSQDDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Otto et al.
Nature chemistry, 4(7), 534-538 (2012-06-22)
Solvents have a profound influence on chemical reactions in solution and have long been used to control their outcome. Such effects are generally considered to be governed by thermodynamics; however, little is known about the steric effects of solvent molecules.
Christian R Evenhuis et al.
The journal of physical chemistry. A, 115(23), 5992-6001 (2011-02-12)
The photodissociation of methyl iodide in the A band is studied by full-dimensional (9D) wave packet dynamics calculations using the multiconfigurational time-dependent Hartree approach. The potential energy surfaces employed are based on the diabatic potentials of Xie et al. [J.
Microhydration effects on the intermediates of the S(N)2 reaction of iodide anion with methyl iodide.
Keisuke Doi et al.
Angewandte Chemie (International ed. in English), 52(16), 4380-4383 (2013-01-31)
Warayuth Sajomsang et al.
International journal of biological macromolecules, 50(1), 263-269 (2011-11-22)
Five water-soluble chitosan derivatives were carried out by quaternizing either iodomethane or N-(3-chloro-2-hydroxypropyl) trimethylammonium chloride (Quat188) as a quaternizing agent under basic condition. The degree of quaternization (DQ) ranged between 28±2% and 90±2%. The antifungal activity was evaluated by using
Kazunori Kawamura et al.
Nuclear medicine and biology, 39(1), 89-99 (2011-08-13)
To explore the possible use of positron emission tomography (PET) probes for imaging of I(2)-imidazoline receptors (I(2)Rs) in peripheral tissues, we labeled two new I(2)R ligands, 2-[2-(o-tolyl)vinyl]-4,5-dihydro-1H-imidazole (K(i) for I(2)Rs, 3.7 nM) and 2-[2-(o-tolyl)ethyl]-4,5-dihydro-1H-imidazole (K(i) for I(2)Rs, 1.7 nM) with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service