Skip to Content
Merck
All Photos(1)

Key Documents

50233-U

Spherisorb® ODS1 HPLC Column

5 μm particle size, L × I.D. 15 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Agency

suitable for USP L1

Quality Level

feature

endcapped

manufacturer/tradename

Spherisorb®

extent of labeling

6.2% Carbon loading

parameter

20-45 °C temp. limit
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

matrix

octadecyl

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

80 Å

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Spherisorb is a registered trademark of Waters

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service