Skip to Content
Merck
All Photos(1)

Documents

49107-U

Supelco

EPA 552.2 Haloacetic Acids Mix

certified reference material, 2000 μg/mL each component in methyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

EPA 552.2

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in methyl tert-butyl ether

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

EPA 552.2 Haloacetic Acids Mix is a multi-component solution containing bromoacetic acid, bromochloroacetic acid, bromodichloroacetic acid, chloroacetic acid, dibromochloroacetic acid, dibromoacetic acid, dichloroacetic acid, tribromoacetic acid, and trichloroacetic acid as certified reference materials (CRMs). The raw materials and solvents used in the preparation of EPA 552.2 Haloacetic Acids Mix have been screened for identity and purity. The mixture is gravimetrically prepared and quantitatively analyzed by gas chromatography (GC).
US EPA 500 Series methods are analytical methods for identifying and quantifying organic contaminants such as volatile organic compounds (VOCs), pesticides, synthetic organic compounds (SOCs), and trihalomethane disinfection byproducts (THMs) in raw source water, finished drinking water, and drinking water at all stages of treatment.

Application

EPA Method 552.2 Haloacetic Acids Mix is used as a CRM to quantify halogenated carbonaceous disinfection by-products (C-DBPs) in drinking water, tap water, treated water, and wastewater samples by gas chromatography (GC) based techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Bromoacetic acid
    Bromochloroacetic acid
    Bromodichloroacetic acid
    Chloroacetic acid
    Dibromochloroacetic acid
    Dibromoacetic acid
    Dichloroacetic acid
    Tribromoacetic acid
    Trichloroacetic acid
    See All (9)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-27.4 °F - closed cup

Flash Point(C)

-33.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Degradation of haloacetic acids with the Fenton-like and analysis by GC-MS: use of bioassays for monitoring of genotoxic, mutagenic and cytotoxic effects
Mour?o AO, et al.
Environmental Monitoring and Assessment, 191(8), 513-513 (2019)
Evidence of arsenic release promoted by disinfection by-products within drinking-water distribution systems
Andra SS, et al.
The Science of the Total Environment, 472(8), 1145-1151 (2014)
Predicting the formation of haloacetonitriles and haloacetamides by simulated distribution system tests
Sfynia C, et al.
Procedia Engineering, 186(8), 186-192 (2017)
Carbonaceous and nitrogenous disinfection byproduct precursor variation during the reversed anaerobic-anoxic-oxic process of a sewage treatment plant
Han H, et al.
Environments, 65(8), 335-346 (2018)
Formation of halogenated C-, N-DBPs from chlor (am) ination and UV irradiation of tyrosine in drinking water
Chu W, et al.
Environmental Pollution (Barking, Essex : 1987), 161, 8-14 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service