48545
Isophorone
analytical standard
Synonym(s):
3,5,5-Trimethyl-2-cyclohexen-1-one
About This Item
grade
analytical standard
vapor density
4.77 (vs air)
vapor pressure
0.2 mmHg ( 20 °C)
autoignition temp.
864 °F
expl. lim.
3.8 %
packaging
ampule of 5000 mg
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
refractive index
n20/D 1.476 (lit.)
bp
213-214 °C (lit.)
mp
−8 °C (lit.)
density
0.923 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
format
neat
storage temp.
2-8°C
SMILES string
CC1=CC(=O)CC(C)(C)C1
InChI
1S/C9H14O/c1-7-4-8(10)6-9(2,3)5-7/h4H,5-6H2,1-3H3
InChI key
HJOVHMDZYOCNQW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
204.8 °F - closed cup
Flash Point(C)
96.0 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service