Skip to Content
Merck
All Photos(1)

Key Documents

48411

Supelco

1,1,2-Trichloro-1,2,2-trifluoroethane

certified reference material, 1000 μg/mL in methanol

Synonym(s):

1,1,2-Trichloro-1,2,2-trifluoroethane solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

InChI

1S/C2Cl3F3/c3-1(4,6)2(5,7)8

InChI key

AJDIZQLSFPQPEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Ozone 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Michael et al.
International journal of pharmaceutics, 221(1-2), 165-174 (2001-06-09)
The nature of the drug-drug aggregation phenomena between salmeterol xinafoate and fluticasone propionate used in a metered-dose inhaler system has been examined. Interactions between the drugs in the solvents 1,1,2-trichlorotrifloroethane (CFC-113) and 1,1,1,2-tetrafluoroethane (HFA-134a) have been characterised using a focused
J A Richt et al.
Medical microbiology and immunology, 182(5), 271-280 (1993-11-01)
Treatment of homogenates from Borna disease virus (BDV)-infected brain tissue or cell cultures with Freon-113 yielded infectious particles with a buoyant density of 1.16-1.22 g/ml. Positive- and negative-stranded BDV-specific RNA species as well as three virus-specific proteins, known to be
M Daubländer et al.
Anesthesia & pain control in dentistry, 1(3), 161-166 (1992-01-01)
The effects of three lidocaine-containing solutions with and without frigen (freon-113) as a propellant on cardiovascular parameters and oxygen saturation, after different waiting periods and with different dosages applied, were investigated in 130 outpatients undergoing dental treatment in the maxilla
S Sousa et al.
The Journal of applied bacteriology, 63(6), 559-564 (1987-12-01)
An initial investigation was made of the bactericidal and sporicidal activity of the dry cleaning solvent Freon-113 (1,1,2-trichlorotrifluoroethane) under various conditions. Representative bacterial strains selected for this study were Escherichia coli (ATCC 11229), Staphylococcus aureus (ATCC 6538) and spores of
J R Akse et al.
Life support & biosphere science : international journal of earth space, 1(3-4), 159-167 (1995-04-01)
The Pd on Al2O3 catalyst used in the projected Space Station's Trace Contaminant Control System (TCCS) catalytic oxidizer can be poisoned by volatile halogen-, sulfur-, and nitrogen-containing organic species. Catalytically Active Regenerable Sorbents (CARS) eliminate these problematic contaminants and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service