Skip to Content
Merck
All Photos(1)

Key Documents

48347

Supelco

Styrene solution

certified reference material, 200 μg/mL in methanol

Synonym(s):

Phenylethylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8
CAS Number:
Molecular Weight:
104.15
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

C=Cc1ccccc1

InChI

1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2

InChI key

PPBRXRYQALVLMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

50.0 °F

Flash Point(C)

10 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Saviello et al.
Analytica chimica acta, 843, 59-72 (2014-08-26)
Synchrotron-based Fourier transform infrared micro-spectroscopy (SR-μFTIR) was used to map photo-oxidative degradation of acrylonitrile-butadiene-styrene (ABS) and to investigate the presence and the migration of additives in historical samples from important Italian design objects. High resolution (3×3 μm(2)) molecular maps were
A comprehensive evaluation of the potential health risks associated with occupational and environmental exposure to styrene.
Joshua T Cohen et al.
Journal of toxicology and environmental health. Part B, Critical reviews, 5(1-2), 1-265 (2002-05-16)
Aisling Mooney et al.
Applied microbiology and biotechnology, 72(1), 1-1 (2006-07-11)
Large quantities of the potentially toxic compound styrene are produced and used annually by the petrochemical and polymer-processing industries. It is as a direct consequence of this that significant volumes of styrene are released into the environment in both the
Günter Speit et al.
Mutation research, 589(1), 67-79 (2005-01-18)
Results from new genotoxicity tests in laboratory animals have necessitated a comprehensive re-evaluation of the mutagenic potential of styrene in vivo. Available data suggest that styrene, after being metabolized to styrene oxide, is weakly positive in indicator tests detecting DNA
Bum Gun Kwon et al.
Environmental pollution (Barking, Essex : 1987), 188, 45-49 (2014-02-21)
Beach sand and seawater taken from the coastlines of the North-East Pacific Ocean and Hawaii State were investigated to determine the causes of global chemical contamination from polystyrene (PS). All samples were found to contain styrene monomer (SM), styrene dimers (SD)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service