Skip to Content
Merck
All Photos(1)

Key Documents

48292

Supelco

Acetophenone solution

certified reference material, 2000 μg/mL in methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O
CAS Number:
Molecular Weight:
120.15
EC Number:
UNSPSC Code:
12000000

grade

certified reference material

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methylene chloride

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

InChI

1S/C8H8O/c1-7(9)8-5-3-2-4-6-8/h2-6H,1H3

InChI key

KWOLFJPFCHCOCG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dejan Vražič et al.
Molecules (Basel, Switzerland), 18(1), 74-96 (2013-01-25)
The Brønsted-acidic ionic liquid 1-methyl-3-(4-sulfobutyl)imidazolium triflate [BMIM(SO(3)H)][OTf] was demonstrated to act efficiently as solvent and catalyst for the halogenation of activated organic compounds with N-halosuccinimides (NXS) under mild conditions with short reaction times. Methyl aryl ketones were converted into α-halo
Raphaël F Guignard et al.
Chemical communications (Cambridge, England), 47(44), 12185-12187 (2011-10-15)
A new concise route to Polycyclic Aromatic Hydrocarbons (PAHs) through radical addition and cyclisation of xanthates is described.
Fang Dong et al.
Journal of plant physiology, 169(3), 217-225 (2012-01-03)
Acetophenone (AP) and 1-phenylethanol (1PE) are the two major endogenous volatile compounds in flowers of Camellia sinensis var. Yabukita. Until now no information has been available on the biosynthesis of AP and 1PE in plants. Here we propose that AP
V Selvarani et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 91, 329-337 (2012-03-14)
Four tetradentate (N(2)O(2)) and tridentate (NO(2)) Schiff base compounds (L1-L4) with propargyl moiety were prepared by the condensation of 1-[2-hydroxy-4-(prop-2-yn-1-yloxy)phenyl]ethanone with various aliphatic amines. The newly synthesized compounds (L1-L4) were characterized on the basis of the results of elemental analysis
Eric R Bittner et al.
The Journal of chemical physics, 137(22), 22A551-22A551 (2012-12-20)
The standard model for molecular recognition of an odorant is that receptor sites discriminate by molecular geometry as evidenced that two chiral molecules may smell very differently. However, recent studies of isotopically labeled olfactants indicate that there may be a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service