Skip to Content
Merck
All Photos(1)

Documents

48264

Supelco

2,3,4,6-Tetrachlorophenol solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Cl4O
CAS Number:
Molecular Weight:
231.89
Beilstein:
779754
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Oc1c(Cl)cc(Cl)c(Cl)c1Cl

InChI

1S/C6H2Cl4O/c7-2-1-3(8)6(11)5(10)4(2)9/h1,11H

InChI key

VGVRPFIJEJYOFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M K Männisto et al.
Water research, 35(10), 2496-2504 (2001-06-08)
The composition and chlorophenol-degrading potential of groundwater bacterial community in a permanently cold, oxygen-deficient chlorophenol contaminated aquifer at Kärkölä, Finland was studied with the aim of evaluating in situ bioremediation potential. The groundwater contained from 10(4) to 10(7) microscopically counted
Pavel Smejtek et al.
The Journal of chemical physics, 120(3), 1383-1394 (2004-07-23)
We studied the effect of segmented solvent molecules on the free energy of transfer of small molecules from water into alkanes (hexane, heptane, octane, decane, dodecane, tetradecane, and hexadecane). For these alkanes we measured partition coefficients of benzene, 3-methylindole (3MI)
J H Langwaldt et al.
Applied microbiology and biotechnology, 49(6), 663-668 (1998-07-31)
Air-lift percolator experiments simulated in situ subsurface degradation of 2,4,6-trichlorophenol, 2,3,4,6-tetrachlorophenol and pentachlorophenol, in mixtures and individually, by indigenous microorganisms from a chlorophenol-contaminated aquifer. Inoculation with a chlorophenol(CP)-degrading gram-positive isolate from the CP-contaminated groundwater did not significantly increase CP degradation
H Huang et al.
Chemosphere, 41(6), 943-951 (2000-06-23)
A kinetic model is developed for PCDD formation from chlorophenol catalysed by incinerator fly ash. The key step in the model is a Langmuir-Hinshelwood type elementary step for the coupling of two adsorbed chlorophenol species to PCDD. Kinetic expression is
J S Karns et al.
Applied and environmental microbiology, 46(5), 1176-1181 (1983-11-01)
Resting cells of 2,4,5-trichlorophenoxyacetic acid-grown Pseudomonas cepacia AC1100 were able to completely and rapidly dechlorinate several chlorine-substituted phenols, including 2,4,5-trichlorophenol, 2,3,4,6-tetrachlorophenol, and pentachlorophenol. Several other trichlorophenols were only partially dechlorinated. The evidence suggests that 2,4,5-trichlorophenol is an intermediate in the

Protocols

Analytical standard separation of various phenols for research and industrial applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service