Skip to Content
Merck
All Photos(1)

Key Documents

48162

Supelco

1-Methylnaphthalene solution

certified reference material, 2000 μg/mL in methanol

Synonym(s):

α-Methylnaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C11H10/c1-9-5-4-7-10-6-2-3-8-11(9)10/h2-8H,1H3

InChI key

QPUYECUOLPXSFR-UHFFFAOYSA-N

General description

1-Methylnaphthalene is an oily liquid having an earthy phenolic type of odor.[1] It is an important constituent of polycyclic aromatic hydrocarbons[2], which can be commonly obtained from coal and petroleum oils.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Encyclopedia of Food and Color Additives, 1 (1997)
Issues in Radiation Biology and Toxicology Research (2013)
Y Murata et al.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie, 44(1), 47-54 (1992-03-01)
Pulmonary alveolar proteinosis was induced at a 100% incidence in B6C3F1 female mice by twice weekly painting the back skin with methylnaphthalene for 30 weeks to give a total dose of 7.14 g/kg b.wt. Semithin sections were used for defining
Mykola Seredych et al.
Langmuir : the ACS journal of surfaces and colloids, 26(1), 227-233 (2009-12-30)
Two synthetic, polymer-derived carbons were modified with urea to incorporate nitrogen surface functional groups. Then they were investigated as adsorbents of dibenzothiophene (DBT) and 4, 6-dimethyldibenzothiophene (DMDBT) from simulated diesel fuel under dynamic conditions with the total concentration of sulfur
U Kulka et al.
Mutation research, 208(3-4), 155-158 (1988-07-01)
Chromosome analyses were carried out in human lymphocytes treated in vitro with 1- and 2-methylnaphthalene (1-MN, 2-MN) in the presence and absence of the mammalian metabolic activation system, S9 mix. Without S9 mix there was no indication of induction of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service