Skip to Content
Merck
All Photos(1)

Key Documents

48097

Supelco

Aroclor 1016 solution

certified reference material, 1000 μg/mL in isooctane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Agency

EPA 8080,8081,8082,Mar ′90, Dec ′90, Aug ′91 Statement of Work
EPA OLM04 (Statement of Work)

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

Clc1cc(cc(c1)c2cc(ccc2)Cl)Cl

InChI

1S/C12H7Cl3/c13-10-3-1-2-8(4-10)9-5-11(14)7-12(15)6-9/h1-7H

InChI key

RIBGNAJQTOXRDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aroclor 1016, a hazardous environmental contaminant,[1] is a complex mixture of polychlorinated biphenyl (PCB) congeners, prepared by the chlorination of biphenyl.[2]

Application

Aroclor 1016 may be used as a certified reference standard for the quantification of the analyte in environmental matrices[3][1] using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A disposable immunomagnetic electrochemical sensor based on functionalised magnetic beads and carbon-based screen-printed electrodes (SPCEs) for the detection of polychlorinated biphenyls (PCBs)
Centi.S, et al.
Analytica Chimica Acta, 538(1-2), 205-212 (2005)
Determination of PCBs in soils/sediments by microwave-assisted extraction and GC/ECD or ELISA
Avila-Lopez V, et al.
Environmental Science & Technology, 29(10), 2709-2712 (1995)
Marika Viisimaa et al.
Environmental technology, 33(16-18), 2041-2048 (2012-12-18)
The potential and feasibility of treating soil contaminated with electrical insulating oil, Aroclor 1016, containing polychlorinated biphenyls (PCBs) with stabilized hydrogen peroxide were evaluated using columns packed with soils of two different matrixes. The column experiments showed that PCBs degraded
Monika G Baldridge et al.
Reproductive toxicology (Elmsford, N.Y.), 17(5), 567-573 (2003-10-14)
Polychlorinated biphenyls (PCBs) are ubiquitous man-made toxicants capable of endocrine disruption. Studies in several species have shown that exposure to PCBs and their hydroxylated metabolites reduces fecundity and decreases circulating concentrations of thyroid hormones, causing serious reproductive and developmental defects.
Activity of environmentally relevant low doses of endocrine disruptors and the bisphenol A controversy: initial results confirmed.
D M Sheehan
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 224(2), 57-60 (2000-05-12)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service