Skip to Content
Merck
All Photos(1)

Key Documents

48067

Supelco

Bromochloromethane solution

certified reference material, 2000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2BrCl
CAS Number:
Molecular Weight:
129.38
UNSPSC Code:
12000000
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Agency

EPA 601,8010,8021 (1987),8021A (1990)

product line

TraceCERT®

feature

standard type surrogate

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/CH2BrCl/c2-1-3/h1H2

InChI key

JPOXNPPZZKNXOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromochloromethane can be produced from dichloromethane, via halogen exchange reaction using either liquid bromine or gaseous hydrogen bromide.[1] It can find applications as a fire extinguishing fluid, especially in aircrafts and other portable extinguishers.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Ozone 1 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ullmann's Fine Chemicals (2014)
Orme-Zavaleta J and Connery J
Drinking Water Health Advisory: Volatile Organic Compounds (1990)
M L Gargas et al.
Toxicology and applied pharmacology, 86(3), 341-352 (1986-12-01)
In vivo metabolic constants were determined in male Fischer rats for five chemicals: 1,1-dichloroethylene (1,1-DCE), diethyl ether (DE), bromochloromethane (BCM), methyl chloroform (MC), and carbon tetrachloride (CCl4). A closed recirculated exposure system was used to collect a series of uptake
Makoto Mitsumori et al.
The British journal of nutrition, 108(3), 482-491 (2011-11-09)
The effects of the anti-methanogenic compound, bromochloromethane (BCM), on rumen microbial fermentation and ecology were examined in vivo. Japanese goats were fed a diet of 50 % Timothy grass and 50 % concentrate and then sequentially adapted to low, mid and high
Gunjan Goel et al.
The British journal of nutrition, 101(10), 1484-1492 (2009-02-27)
Bromochloromethane (BCM), a halogenated methane analogue, was evaluated for its anti-methanogenic activity in both batch and continuous fermentation systems. For batch fermentation, a roughage-based substrate was incubated with mixed rumen microflora for 24 h at two concentrations of BCM (5

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service