Skip to Content
Merck
All Photos(1)

Key Documents

47641-U

Supelco

Reversed Phase Test Mix 2

certified reference material, in acetonitrile: water (varied conc.), solution

Synonym(s):

HPLC Test Mixture for Reversed Phase Columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

product line

TraceCERT®

form

solution

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

in acetonitrile: water (varied conc.)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

compatibility

use to QA hydrophobic reversed phase columns

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    N,N-Diethyl-m-toluamide 600 μg/mL
    Phenol 700 μg/mL
    Toluene 4000 μg/mL
    Uracil 5 μg/mL

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

45.0 °F - closed cup

Flash Point(C)

7.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service