47548-U
EPA 8270 GC-MS Tuning Solution II
certified reference material, 1000 μg/mL each component in dichloromethane
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
Agency
EPA 8270
product line
TraceCERT®
CofA
current certificate can be downloaded
feature
standard type tuning solution
packaging
ampule of 1 mL
concentration
1000 μg/mL each component in dichloromethane
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
multi-component solution
storage temp.
2-8°C
Application
Other Notes
Legal Information
Analyte
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Carc. 1A - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service