Skip to Content
Merck
All Photos(1)

Key Documents

47509-U

Supelco

Benzyl alcohol solution

certified reference material, 2000 μg/mL in methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methylene chloride

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luis E Santos-Figueroa et al.
Chemical Society reviews, 42(8), 3489-3613 (2013-02-13)
This review focuses on examples reported in the years 2010-2011 dealing with the design of chromogenic and fluorogenic chemosensors or reagents for anions.
Kelly Roberta Francisco et al.
Langmuir : the ACS journal of surfaces and colloids, 28(41), 14531-14539 (2012-09-13)
Polymers formed by the self-assembly of a bis(urea)-based polymer, 2,4-bis(2-ethylhexylureido)toluene (EHUT), in organic solvents such as octane are promising systems with remarkable rheological properties. This is the first self-assembled polymer recently reported as a hydrodynamic drag reducer for hydrocarbons. The
Mon-Wei Hsiao et al.
Dalton transactions (Cambridge, England : 2003), 42(6), 2041-2051 (2012-11-28)
A series of calcium complexes supported by NNO-tridentate ketiminate ligands have been synthesized and characterized. X-ray structural studies indicate that these complexes bear a mononuclear feature with hexa-coordinated calcium centers bonding to two ketiminate ligands. Polymerization of L-lactide catalyzed by
Aaron L Acton et al.
Biomacromolecules, 14(2), 564-574 (2013-01-12)
The increasing use of drug combinations to treat disease states, such as cancer, calls for improved delivery systems that are able to deliver multiple agents. Herein, we report a series of novel Janus dendrimers with potential for use in combination
Pierre-Yves Blanchard et al.
Langmuir : the ACS journal of surfaces and colloids, 28(38), 13741-13745 (2012-09-06)
Electrocatalytic behavior of TEMPO derivative SAMs on gold has been studied in the presence of benzyl alcohol. The results demonstrate that interfacial activity of the SAMs can be enhanced by diluting the TEMPO moiety with an alkyl passive matrix. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service