47444-U
EPA 625 Semivolatile Calibration Kit
analytical standard
About This Item
grade
analytical standard
Agency
EPA 625
CofA
current certificate can be downloaded
feature
standard type calibration
packaging
pkg of 1 kit
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
multi-component solution
storage temp.
-10 to -25°C
General description
Packed GC columns are described in the EPA 625 method, but chromatographic conditions can be changed (e.g., capillary columns may be used) if the changes do not make the method "less accurate or less precise" than the standard method.
Application
Kit Components Only
- 625 Semivolatile Calibration Mix (506559) 1 mL
- TCL Benzidines Mix (48906) 1 mL
- N-Nitrosodiphenylamine solution (40060) 1 mL
Signal Word
Danger
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 1 - STOT SE 3
Target Organs
Blood, Central nervous system, Eyes,Central nervous system, Respiratory system
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service