Skip to Content
Merck
All Photos(1)

Documents

47408

Supelco

EPA VOC Mix 6

2000 μg/mL each component in methanol, analytical standard

Synonym(s):

Volatile Organic Compounds Mix 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

Quality Level

Agency

EPA 8260
suitable for EPA 8260

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 4 × 1 mL

concentration

2000 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

EPA VOC mix 6 is a standard calibration mix, which may be used for the analysis of volatile organic compounds by gas-chromatography coupled with mass spectrometry technique (GC-MS).

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Bromomethane
    Chloroethane
    Chloromethane
    Dichlorodifluoromethane
    Trichlorofluoromethane
    Vinyl chloride

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1A - Flam. Liq. 2 - Ozone 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service