Skip to Content
Merck
All Photos(1)

Key Documents

47260-U

Supelco

EPA 8270 Acids Surrogate Spike Mix HC

certified reference material, 10 mg/mL each component in methanol, ampule of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 8270

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type surrogate

packaging

ampule of 1 mL

concentration

10 mg/mL each component in methanol

technique(s)

gas chromatography (GC): suitable

format

multi-component solution

storage temp.

2-30°C

Other Notes

This certified reference material may be used to identify and quantitate key fatty acid methyl esters in a variety of food products. The complex nature of this mix eliminates the need to purchase multiple mixtures while minimizing expenses.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    2-Fluorophenol
    Phenol-d6
    2,4,6-Tribromophenol

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service