Recommended Products
grade
analytical standard
Agency
ASTM® D6729,D6730,D6733
packaging
ampule of 0.1 mL
concentration
(typical values shown)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
petroleum
format
multi-component solution
storage temp.
2-8°C
General description
P-I-A-N-O Mix to used determine retention times and indices and monitor response factors for components of complex mixtures of hydrocarbons. These standards are complex mixes of known quantities of hydrocarbons, accurately prepared by weight to three decimal places.
Application
P-I-A-N-O Mix may be used as an analytical standard for the identification and quantification of gaseous organic compounds emitted from biomass using two-dimensional gas chromatography coupled to time of flight mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Legal Information
ASTM is a registered trademark of American Society for Testing and Materials
Analyte
Description
n-Paraffins 18.9 % (w/w)
Isoparaffins 18.8 % (w/w)
Aromatics 23.3 % (w/w)
Naphthenes 20.5 % (w/w)
Olefins 18.5 % (w/w)
analytical column
Product No.
Description
Pricing
standard
Product No.
Description
Pricing
Flash Point(F)
closed cup
Flash Point(C)
closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Identification and quantification of gaseous organic compounds emitted from biomass burning using two-dimensional gas chromatography-time-of-flight mass spectrometry
Hatch.EL, et al.
Atmospheric Chemistry and Physics, 15(4), 1865-1899 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service