Skip to Content
Merck
All Photos(1)

Key Documents

442748

Supelco

Permethrin (isomers)

PESTANAL®, analytical standard

Synonym(s):

Permethrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20Cl2O3
CAS Number:
Molecular Weight:
391.29
Beilstein:
5765325
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

-10 to -25°C

SMILES string

CC1(C)C(\C=C(/Cl)Cl)C1C(=O)OCc2cccc(Oc3ccccc3)c2

InChI

1S/C21H20Cl2O3/c1-21(2)17(12-18(22)23)19(21)20(24)25-13-14-7-6-10-16(11-14)26-15-8-4-3-5-9-15/h3-12,17,19H,13H2,1-2H3

InChI key

RLLPVAHGXHCWKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is being discontinued but product 45614-250mg may be an acceptable replacement.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Analysis of permethrin isomers in composite diet samples by molecularly imprinted solid-phase extraction and isotope dilution gas chromatography-ion trap mass spectrometry.
Vonderheide AP, et al.
Journal of Chromatography A, 1216(22), 4633-4640 (2009)
An ecofriendly and stability-indicating HPLC method for determination of permethrin isomers: application to pharmaceutical analysis.
Afshar M, et al.
Journal of Chemistry, 2013 (2012)
Elimination and metabolism of permethrin isomers in rainbow trout.
Glickman AH, et al.
Toxicology and Applied Pharmacology, 57(1), 88-98 (1981)
Marie Varloud et al.
Parasitology research, 114(3), 965-973 (2014-12-31)
This study was designed to compare the efficacy of three topical combinations on dogs in outdoor conditions against adult cat fleas (Ctenocephalides felis), flea egg hatch and emergence, and against adult brown dog ticks (Rhipicephalus sanguineus sensu lato). Treatment was
L Fuortes
Veterinary and human toxicology, 41(2), 92-93 (1999-04-07)
Although naturally derived pyrethrins are well recognized sensitizers, there is currently some degree of controversy regarding the sensitizing potential of synthetic pyrethroids. The literature is briefly reviewed, and an outbreak of several cases of urticaria occurring in children from 1

Articles

Extraction of Permethrin Pesticides from Spinach Using QuEChERS Methodology with Automated Shaking

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service