Skip to Content
Merck
All Photos(1)

Key Documents

442740

Supelco

Pentachloroethane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CCl3CHCl2
CAS Number:
Molecular Weight:
202.29
Beilstein:
1736845
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.502 (lit.)

bp

161-162 °C (lit.)

density

1.68 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

room temp

SMILES string

ClC(Cl)C(Cl)(Cl)Cl

InChI

1S/C2HCl5/c3-1(4)2(5,6)7/h1H

InChI key

BNIXVQGCZULYKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J W Nichols et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 31(2), 229-242 (1996-06-01)
A physiologically based toxicokinetic model was developed to describe dermal absorption of waterborne organic chemicals by fish. The skin was modeled as a discrete compartment into which compounds diffuse as a function of chemical permeability and the concentration gradient. The
G Bronzetti et al.
Teratogenesis, carcinogenesis, and mutagenesis, 9(6), 349-357 (1989-01-01)
Tetrachloroethane (TTCE), pentachloroethane (PCE), and hexachloroethane (HCE) were tested in diploid strain (D7) of the yeast Saccharomyces cerevisiae in suspension test with and without mammalian metabolic activation (S9). TTCE, PCE, and HCE gave positive results on cells harvested from logarithmic
A heme model system for the reduction of substrates by microsomal cytochrome P 450.
D Mansuy et al.
Biochemical and biophysical research communications, 104(4), 1651-1657 (1982-02-26)
Ryo Iwakiri et al.
Bioscience, biotechnology, and biochemistry, 68(6), 1353-1356 (2004-06-25)
We engineered biphenyl-degrading Alcaligenes sp. strain KF711 for total degradation of pentachloroethane (PCA), which expresses a modified camphor monooxygenase and a hybrid dioxygenase consisting of TodC1 (a large subunit of toluene dioxygenase of Pseudomonas putida F1) and BphA2-BphA3-pbhA4 (a small
J Nichols et al.
Environmental health perspectives, 102(11), 952-956 (1994-11-01)
Outputs from a physiologically based toxicokinetic (PB-TK) model for fish were visualized by mapping time-series data for specific tissues onto a three-dimensional representation of a rainbow trout. The trout representation was generated in stepwise fashion: 1) cross-section images were obtained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service