Skip to Content
Merck
All Photos(1)

Key Documents

442506

Supelco

Carbazole

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9N
CAS Number:
Molecular Weight:
167.21
Beilstein:
3956
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor pressure

400 mmHg ( 323 °C)

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

355 °C (lit.)

mp

243-246 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

c1ccc2c(c1)[nH]c3ccccc23

InChI

1S/C12H9N/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8,13H

InChI key

UJOBWOGCFQCDNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carbazoles belong to the family of polycyclic aromatic hydrocarbons (PAHs) compounds.

Application

Carbazole may be used as a reference standard for the determination of carbazole in:
  • e-cigarette aerosols by thermal desorption-gas chromatography-flame ionization detection-time of flight mass spectrometry (TD-GC-FID-TOFMS).
  • Organic gunshot residue samples and in unfractionated coal tar samples by TD-GC-MS working in selected ion monitoring (SIM) mode
  • Soil samples affected by tire fires by comprehensive two-dimensional gas chromatography (GC x GC)coupled to TOFMS operating on electron impact (EI) ionization mode.
  • Sediment extracts by high performance liquid chromatography in conjunction with diode array detection (HPLC-DAD) and GC-MS in SIM mode.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 2 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

428.0 °F - closed cup

Flash Point(C)

220.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Speciation of nitrogen-containing compounds in an unfractionated coal tar sample by comprehensive two-dimensional gas chromatography coupled to time-of-flight mass spectrometry.
da Silva JM, et al.
Journal of Chromatography A, 1373, 159-168 (2014)
Chemical characterisation of aerosols emitted by electronic cigarettes using thermal desorption-gas chromatography-time of flight mass spectrometry.
Rawlinson C, et al.
Journal of Chromatography A, 1497, 144-154 (2017)
Initial evaluation of inlet thermal desorption GC-MS analysis for organic gunshot residue collected from the hands of known shooters.
Stevens B, et al.
Forensic Chemistry, 2, 55-62 (2016)
Characterization of organic aromatic compounds in soils affected by an uncontrolled tire landfill fire through the use of comprehensive two-dimensional gas chromatography-time-of-flight mass spectrometry
Escobar-Arnanz J, et al.
Journal of Chromatography A, 1536, 163-175 (2018)
Automated fractionation procedure for polycyclic aromatic compounds in sediment extracts on three coupled normal-phase high-performance liquid chromatography columns
Lubcke-von Varel U, et al.
Journal of Chromatography A, 1185(1), 31-42 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service