Skip to Content
Merck
All Photos(1)

Key Documents

42439

Supelco

Myristic acid-1,2,3,4-13C4

analytical standard

Synonym(s):

Tetradecanoic-1,2,3,4-13C4 acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$704.00

$704.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$704.00

About This Item

Empirical Formula (Hill Notation):
13C4C10H28O2
Molecular Weight:
232.34
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.24

$704.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

extent of labeling

≥99%

storage temp.

2-8°C

SMILES string

CCCCCCCCCC[13CH2][13CH2][13CH2][13C](O)=O

InChI key

TUNFSRHWOTWDNC-AUHOWUHASA-N

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service