Skip to Content
Merck
All Photos(1)

Key Documents

41934

Supelco

7-Hydroxyflavone

analytical standard

Synonym(s):

7-Hydroxy-2-phenyl-4H-1-benzopyran-4-one, NSC 94258

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$142.00

$142.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$142.00

About This Item

Empirical Formula (Hill Notation):
C15H10O3
CAS Number:
Molecular Weight:
238.24
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$142.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

245-247 °C (lit.)

SMILES string

Oc1ccc2C(=O)C=C(Oc2c1)c3ccccc3

InChI

1S/C15H10O3/c16-11-6-7-12-13(17)9-14(18-15(12)8-11)10-4-2-1-3-5-10/h1-9,16H

InChI key

MQGPSCMMNJKMHQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant:
  • Acting as a biologically valuable acceptor in O-glycosidation reactions
  • Involved in synthesis of fully phosphorylated flavones for use as pancreatic cholesterol esterase inhibitors
  • For O-methylation with di-Me carbonate
  • Linked by a polymethylene chain for synthesis of α1-adrenoceptor antagonists
  • Involved in Baylis-Hillman reactions
  • Involved in phase-transfer catalyzed glucosylation for synthesis of glucosylated flavonoids

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service