Skip to Content
Merck
All Photos(1)

Key Documents

41917

Supelco

Isobornyl acetate

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$98.40

$98.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$98.40

About This Item

Empirical Formula (Hill Notation):
C12H20O2
CAS Number:
Molecular Weight:
196.29
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.24

$98.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥94.0% (GC)

optical activity

[α]/D -2.0 to 2.0, neat

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.462-1.465
n20/D 1.4635 (lit.)

bp

229-233 °C (lit.)

density

0.983 g/mL at 25 °C (lit.)

SMILES string

CC(=O)O[C@H]1C[C@@H]2CC[C@@]1(C)C2(C)C

InChI

1S/C12H20O2/c1-8(13)14-10-7-9-5-6-12(10,4)11(9,2)3/h9-10H,5-7H2,1-4H3/t9-,10-,12+/m0/s1

InChI key

KGEKLUUHTZCSIP-JBLDHEPKSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service