Skip to Content
Merck
All Photos(1)

Key Documents

41577

Supelco

7-Ethoxycoumarin

analytical standard

Synonym(s):

7-Ethoxy-1-benzopyran-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$123.00

$123.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$123.00

About This Item

Empirical Formula (Hill Notation):
C11H10O3
CAS Number:
Molecular Weight:
190.20
Beilstein:
146200
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$123.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

88-90 °C (lit.)

SMILES string

CCOc1ccc2C=CC(=O)Oc2c1

InChI

1S/C11H10O3/c1-2-13-9-5-3-8-4-6-11(12)14-10(8)7-9/h3-7H,2H2,1H3

InChI key

LIFAQMGORKPVDH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Substrate for CYP2B6

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service