Skip to Content
Merck
All Photos(1)

Key Documents

40302

Supelco

2,6-Dichlorophenol solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Cl2O
CAS Number:
Molecular Weight:
163.00
EC Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C6H4Cl2O/c7-4-2-1-3-5(8)6(4)9/h1-3,9H

InChI key

HOLHYSJJBXSLMV-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N V Azarkina et al.
Biochemistry. Biokhimiia, 75(1), 50-62 (2010-03-25)
In this work, high DeltamicroH+-dependent succinate oxidase activity has been demonstrated for the first time with membrane vesicles isolated from Bacillus subtilis. The maximal specific rate of succinate oxidation by coupled inside-out membrane vesicles isolated from a B. subtilis strain
Lígia Miranda Ferreira Borges et al.
Veterinary parasitology, 147(1-2), 155-160 (2007-04-27)
This study was carried out with the objective of evaluating the efficacy of a 2,6-dichlorophenol (2,6-DCP) lure to control Dermacentor nitens (Acari: Ixodidae). Slow-release formulations of the pheromone formulated with and without cypermethrin were prepared. Olfactometer bioassays were used to
Davide Vione et al.
Chemosphere, 69(10), 1548-1554 (2007-07-10)
2,4-Dichlorophenol (2,4-DCP) and 2,6-dichlorophenol (2,6-DCP) undergo oxidation, nitrosation and nitration in the presence of nitrate under UV irradiation. Nitration is favoured under acidic conditions, most likely because HNO(2) is formed on nitrate photolysis. The most likely photonitration pathway is the
Lígia M F Borges et al.
Experimental & applied acarology, 27(3), 223-230 (2003-02-21)
The objective of this study was to evaluate the role of 2,6-dichlorophenol (2,6-DCP) as sex pheromone of Anocentor nitens. Sex pheromones were extracted by submerging 150 female fed for 6 days in hexane (3 ml) for two hours and sonicating
Yu Tian et al.
Water research, 47(1), 279-288 (2012-11-06)
Distribution variation of a metabolic uncoupler, 2,6-dichlorophenol (2,6-DCP), in long-term sludge culture was studied, and the effects on sludge reduction and biological inhibition of this chemical during the 90-day operation were established. The extracellular polymeric substance (EPS) matrix functioned as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service