Skip to Content
Merck
All Photos(1)

Key Documents

40252-U

Supelco

4-Methylphenol solution

certified reference material, 5000 μg/mL in methanol

Synonym(s):

4-Methylphenol solution, p-Cresol solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Agency

EPA TO-8
NIOSH 2001
OSHA 32

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Oc1ccc(cc1)C

InChI

1S/C7H8O/c1-6-2-4-7(8)5-3-6/h2-5,8H,1H3

InChI key

IWDCLRJOBJJRNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sean A Newmister et al.
Biochemistry, 51(43), 8571-8582 (2012-10-09)
Cobamides (Cbas) are cobalt (Co) containing tetrapyrrole-derivatives involved in enzyme-catalyzed carbon skeleton rearrangements, methyl-group transfers, and reductive dehalogenation. The biosynthesis of cobamides is complex and is only performed by some bacteria and achaea. Cobamides have an upper (Coβ) ligand (5'-deoxyadenosyl
Aikaterini Papazi et al.
PloS one, 7(12), e51852-e51852 (2012-12-20)
Cultures from the unicellular green alga Scenedesmus obliquus biodegrade the toxic p-cresol (4-methylphenol) and use it as alternative carbon/energy source. The biodegradation procedure of p-cresol seems to be a two-step process. HPLC analyses indicate that the split of the methyl
S-T Huang et al.
Transplantation proceedings, 44(3), 621-624 (2012-04-10)
Uremic toxins are considered cardiovascular and mortality risk factors in chronic kidney disease (CKD) patients. Both p-cresol and indoxyl sulfate have been shown to induce oxidative stress in vitro and subsequent endothelial dysfunction in uremic patients. Our study evaluated the
Jin-zhou Zhu et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 27(12), 4323-4330 (2012-09-11)
Patients afflicted with chronic kidney disease (CKD) typically suffer from cardiovascular disease (CVD) which is a leading cause of patient mortality. It has been demonstrated that two distinct physiological events contribute to this disease state. These include the abundance of
Gunaranjan Paturi et al.
Journal of food science, 77(10), H216-H223 (2012-09-07)
The effects of red meat consumption with and without fermentable carbohydrates on indices of large bowel health in rats were examined. Sprague-Dawley rats were fed cellulose, potato fiber, or potato-resistant starch diets containing 12% casein for 2 wk, then similar

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service