Skip to Content
Merck
All Photos(1)

Key Documents

36831

Supelco

Isocarbophos

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

O-Methyl O-[2-(isopropoxycarbonyl)phenyl] phosphoramidothioate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$114.00

$114.00


Estimated to ship onMarch 27, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$114.00

About This Item

Empirical Formula (Hill Notation):
C11H16NO4PS
CAS Number:
Molecular Weight:
289.29
Beilstein:
2946521
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$114.00


Estimated to ship onMarch 27, 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

−20°C

SMILES string

COP(N)(=S)Oc1ccccc1C(=O)OC(C)C

InChI

1S/C11H16NO4PS/c1-8(2)15-11(13)9-6-4-5-7-10(9)16-17(12,18)14-3/h4-8H,1-3H3,(H2,12,18)

InChI key

YFVOXLJXJBQDEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Isocarbophos is an organophosphorus pesticide that belongs to the phosphoramidate group of compounds. It is widely used to control chewing and sucking insects and spider mites in cotton, rice, fruit trees, and vegetables. Isocarbophos is a potent acetylcholinesterase (AChE) inhibitor that gets activated by both contact and ingestion, blocking hydrolysis of acetylcholine and resulting in hyperexcitation.
Isocarbophos is not approved for use in European Union (EU) as per Regulation (EC) 1107/2009 (repealing Directive 91/414/EEC).

The EU Commission Regulation and US Environmental Protection Agency specified the maximum residue level (MRL) of isocarbophos in agricultural products as 0.01 mg kg−1 and 1 mg kg−1; and Chinese Agriculture has defined an MRL of 0.05 mg kg−1 for its presence in cereals, fruits, and vegetables.

Isocarbophos has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Isocarbophos CRM may also find its use as described below:

  • To assess the bioaccumulation, metabolic disruption, and oxidative stress in adult zebrafish after exposure to isocarbophos using a 1H NMR-based untargeted metabolomic approach and LC-MS/MS
  • To develop an aptamer-based sensitive lateral flow biosensor (LFB) for rapid determination of isocarbophos pesticide in foods
    To develop an HPLC method for the analysis of dichlorvos, isocarbophos, and methyl parathion in plant leaves using chemiluminescence detection
  • Magnetic solid phase extraction based on magnetite/reduced graphene oxide (MRGO) for the determination of trace isocarbophos residues in different matrices, followed by gas chromatography (GC) detection
  • To develop a dual-modal sensor combined with gold nanoparticles (AuNPs) based colorimetry and persistent luminescence nanorods (PLNRs) based phosphorescence for highly sensitive detection of isocarbophos in vegetables

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service