33305
Mininert® Valve
for use with 20 mm crimp seal, pkg of 12 ea
About This Item
Recommended Products
material
white PTFE valve
packaging
pkg of 12 ea
compatibility
for use with 20 mm crimp seal
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
1. Push the green button to open the valve
2. Insert the needle through the septum and take sample
3. Remove the needle and push to close
Benefits include:
- PTFE construction for inertness. PTFE is useful when working when acids and organic solvents
- Good sealing ability for containing most volalite liquids and gases at low pressures
- Mininert valves can be used at temperatures up to 105°F
- 0.036 in. (0.9 mm) opening accepts up to 20-gauge needles
- Silicone septa
- Mininert Valves for Crimp Top Vials
- Fit 5-100 mL vials (20 mm seal)
- Install without tools
- Inert PTFE construction
Application
They are the ideal closure for the following applications:
- Calibration standards
- Air- or moisture-sensitive fluids
- Derivatizating reagents
- Volatile chemicals
Legal Information
related product
replacement part
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service