Skip to Content
Merck
All Photos(1)

Key Documents

33156-U

Supelco

TMSI+PYRIDINE, 1:4 (Sylon TP)

Synonym(s):

1-(Trimethylsilyl)imidazole - Pyridine mixture, Silylating mixture VII, TMSI+Pyridine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

packaging

pkg of 1 × 25 mL

concentration

(TMSI+Pyridine, 1:4)

SMILES string

c1ccncc1.C[Si](C)(C)n2ccnc2

InChI

1S/C6H12N2Si.C5H5N/c1-9(2,3)8-5-4-7-6-8;1-2-4-6-5-3-1/h4-6H,1-3H3;1-5H

InChI key

PFVKFOIOKNIZKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

TMSI+PYRIDINE, 1:4 (Sylon TP) may be used as a derivatization reagent for the measurement of liver and plasma precursor sterols using gas chromatography coupled with mass spectrometry while investigating the dual-action glycogenolysis/cholesterolgenesis inhibitors that act through inhibition of both glycogen phosphorylase and CYP51.

Legal Information

Sylon is a trademark of Sigma-Aldrich Co. LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison between Solvent Extraction Using Gas Chromatography Mass Spectrometry Triple Quadrupole (GCMS-QQQ) of Cholesterol and Cholesterol Oxidation Derivatives
A.R. Alina
Middle-East Journal of Scientific Research 13, 13, 43-48 (2013)
H James Harwood et al.
Journal of lipid research, 46(3), 547-563 (2004-12-18)
Diabetic dyslipidemia requires simultaneous treatment with hypoglycemic agents and lipid-modulating drugs. We recently described glycogen phosphorylase inhibitors that reduce glycogenolysis in cells and lower plasma glucose in ob/ob mice (J. Med. Chem., 41: 2934, 1998). In evaluating the series prototype

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service