Skip to Content
Merck
All Photos(1)

Key Documents

33046

Supelco

HMDS+TMCS

3:1, pkg of 20 × 1 mL

Synonym(s):

Chlorotrimethylsilane, Trimethylsilylchloride, 1,1,1,3,3,3–Hexamethyldisilazane+Trimethylchlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

grade

for GC derivatization

description

silylation reagent

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

reagent type: derivatization reagent
reaction type: Silylations

packaging

pkg of 20 × 1 mL

concentration

(3:1)

General description

HMDS+TMCS is a derivatizing agent.

Application

It was used for silylation reaction of the sample, during determination of aldoses and ketoses using GC-MS analysis.[1]

Other Notes

Reagent for N-acetyl, O-trimethylesilyl, trimethylsilyl and trimethylsilyl ester.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-0.0 °F - closed cup

Flash Point(C)

-17.78 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatographic determination of campesterol, beta-sitosterol and stigmasterol.
A Rozanski
Analytical chemistry, 38(1), 36-40 (1966-01-01)
T.Luukaninen et al.
Biochimica et Biophysica Acta, 52, 599-599 (1961)
Fangting Ye et al.
Phytochemical analysis : PCA, 17(6), 379-383 (2006-12-06)
A method has been established by which to determine aldoses and ketoses in plant material simultaneously. Monosaccharides were extracted by sonication with 80% ethanol and sugar oximes formed by treatment of the resultant extract with hydroxylamine and pyridine at 90
M Hamberg et al.
Proceedings of the National Academy of Sciences of the United States of America, 71(10), 3824-3828 (1974-10-01)
Methods were developed for quantitative determination of the three major metabolites of arachidonic acid in human platelets, i.e., 12L-hydroxy-5,8,10,14-eicosatetraenoic acid (HETE), 12L-hydroxy-5,8,10-heptadecatrienoic acid (HHT) and 8-(1-hydroxy-3-oxopropyl)-9,12L-dihydroxy-5,10-heptadecadienoic acid (PHD). Aggregation of washed platelets by thrombin was accompanied by release of 1163-2175
J P Kamerling et al.
Carbohydrate research, 41, 7-17 (1975-05-01)
A number of O-acetylated N-acylneuraminic acids, isolated from submandibular glands of cow and horse and from horse erythrocytes, have been characterized by mass spectrometry. On the basis of the typical fragmentation patterns of the pertrimethylsilyl derivatives of the methyl esters

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service