Skip to Content
Merck
All Photos(1)

Key Documents

24636

Supelco

Battery for Vacuum Sampling Pump

for use with Model 1062, 1063, & 1067

Synonym(s):

Battery for Field Sampling Pump

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008

technique(s)

whole air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with Model 1062, 1063, & 1067

Looking for similar products? Visit Product Comparison Guide

General description

Battery for Vacuum Sampling Pump is a DC battery employed with a 10-liter vacuum bag sampler. Once it is adjusted to 12 volts, the bag sampler fills or empties a 10-liter bag in two minutes by applying a vacuum or pressure to the outside of the sample bag. The automatic shut-off switch prevents the sample bag from over-filling.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service