Key Documents
22941
4-Port Sampling and Switching Valve
1/16 in. fitting, maximum temperature 300 °C
About This Item
UNSPSC Code:
41112500
eCl@ss:
32060101
Pricing and availability is not currently available.
material
(Nitronic 60)
feature
number of ports 4
manufacturer/tradename
C4T
VICI Valco®
maximum temperature
300 °C
fitting
1/16 in. fitting
General description
Isolate a column to prevent a compound, emerging from an upstream column, from being irreversibly adsorbed. In the figure, CO2 is separated from air in Column 1, and bypasses Column 2 when the valve is switched to position B. After the CO2 is detected, the valve is switched back to position A and O2, N2, CH4, and CO elute from Column 2 to the detector.
Sampling valve basically supports the discrete volumes of the treated samples stream to periodically inject into the carrier gas stream of the chromatograph. Sampling valves are fitted with electric or pneumatic actuators which help them to be operated automatically.[1] Switching valve are critical if binary pumps are used. It allows employment of different desired mobile phases. Switching valve consists of five parts: control unit, connecting device, valve body, a rotor and preloaded assembly.[2]
Application
Sampling valve may be used to connect the LC and GC instruments to allow fast and dependable introduction of large sample volumes onto a capillary GC column.[3]
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
On-line combination of liquid chromatography and capillary gas chromatography. Preconcentration and analysis of organic compounds in aqueous samples.
Noy, Th, et al.
Journal of High Resolution Chromatography, 11, 181-186 (1988)
Clément Vinauger et al.
Current biology : CB, 28(3), 333-344 (2018-02-06)
How mosquitoes determine which individuals to bite has important epidemiological consequences. This choice is not random; most mosquitoes specialize in one or a few vertebrate host species, and some individuals in a host population are preferred over others. Mosquitoes will
Walt Boyes
Instrumentation Reference Book, 414-414 (2009)
Satinder Ahuja
Chiral Separation Methods for Pharmaceutical and Biotechnological Products, 218-218 (2011)