Skip to Content
Merck
All Photos(1)

Key Documents

22298-U

Precision Sampling syringe needles

volume 25-10000 μL, for use with VICI Series A-2, C, D, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2.0 in. × 0.028 in. × 0.005 in.

Synonym(s):

REPLACEMENT TAPER NEEDLEA-2, C, D PK3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

material

bevel tip stainless steel needle (point style 2)

description

A-2, C, D

feature

point style Beveled point, open end (25-100 μL)

packaging

pkg of 3 ea

manufacturer/tradename

VICI 943050

needle L × O.D. × I.D.

2.0 in. × 0.028 in. × 0.005 in.

volume

25-10000 μL

compatibility

for use with VICI Series A-2, C, D

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service